Dichloro(diethylamino)phosphine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Diethylphosphoramidous dichloride, (Diethylamino)phosphonous Dichloride
H25934-06
H25934-14
H25934-22
AAH25934-06EA
73.8
USD
AAH25934-06
AAH25934-14
AAH25934-22
Dichloro(diethylamino)phosphine 97%
Dichloro(diethylamino)phosphine
Purity: 97%
CAS No.: 1069-08-5
Molecular Formula: C4H10Cl2NP
Formula Weight: 174.01
Flash Point: 83°(181°F)
Refractive Index: 1.497
Odor: Irritating
Boiling Point: 179°
UN No.: 3265
MDL No.: MFCD00013614
Formula:
C₄H₁₀Cl₂NP MW: 174.00 g/mol Boiling Pt: 179 °C Density: 1.196 Flash Pt: 83 °C(181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013614 CAS Number: 1069-08-5 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...