Methyl-3-chloro-4-methylbenzoate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-chloro-4-methylbenzoate

H25933-14 H25933-06
AAH25933-14EA 304 USD
AAH25933-14 AAH25933-06 AAH25933-03
Methyl-3-chloro-4-methylbenzoate 95%
Methyl 3-chloro-4-methylbenzoate

Purity: 95% CAS No.: 56525-63-4 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Melting Point: 27-29° Boiling Point: 119-120°/10mm MDL No.: MFCD00205141

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 119…120 °C/10 mm
Melting Pt: 27…29 °C
Storage Temperature: Ambient
MDL Number: MFCD00205141
CAS Number: 56525-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR