Potassium 2-phenylethyltrifluoroborate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium Trifluoro(phenethyl)borate, Potassium Trifluoro(2-phenylethyl)borate, Potassium Phenethyltrifluoroborate

H25930-03 H25930-06
AAH25930-03EA 56.1 USD
AAH25930-03 AAH25930-06
Potassium 2-phenylethyltrifluoroborate 98%
Potassium 2-phenylethyltrifluoroborate

Purity: 98% CAS No.: 329976-74-1 Molecular Formula: C8H9BF3K Formula Weight: 212.06 Melting Point: 173-175° MDL No.: MFCD09039257

Formula: C₈H₉BF₃K
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD09039257
CAS Number: 329976-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR