Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl ethoxymethylene-3-oxo-4,4,4-trifluorobutyrate, 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylic Acid Ethyl Ester, Ethyl 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylate, 2-(Ethoxymethylene)-4,4,4-trifluoro-3-oxobutyric Acid Ethyl Ester
H25927-18
H25927-09
AAH25927-18EA
428
USD
AAH25927-18
AAH25927-09
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 97%
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate
Purity: 97%
CAS No.: 571-55-1
Molecular Formula: C9H11F3O4
Formula Weight: 240.18
Flash Point: 104°(220°F)
Refractive Index: 1.4290
Boiling Point: 80-82°/1mm
MDL No.: MFCD02677683
Formula:
C₉H₁₁F₃O₄ MW: 240.18 g/mol Boiling Pt: 80…82 °C/1 mm Density: 1.235 Flash Pt: 104 °C(220 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02677683 CAS Number: 571-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...