Albendazole ≥98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: [5-(Propylthio)benzimidazol-2-yl]carbamic Acid Methyl Ester, Methyl [5-(Propylthio)benzimidazol-2-yl]carbamate

H25925-09 H25925-22 H25925-18
AAH25925-09EA 77.7 USD
AAH25925-09 AAH25925-22 AAH25925-18
Albendazole ≥98%
Albendazole

Purity: 98+% CAS No.: 54965-21-8 Molecular Formula: C12H15N3O2S Formula Weight: 265.34 Melting Point: 208-210° MDL No.: MFCD00083232

Formula: C₁₂H₁₅N₃O₂S
MW: 265.33 g/mol
Melting Pt: 208…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00083232
CAS Number: 54965-21-8
EINECS: 259-414-7
Merck Index: 14,210

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR