D-Phenylalanine methyl ester hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-Methyl 2-amino-3-phenylpropanoate hydrochloride, H-D-Phe-OMe.HCl, H-D-Phe-OMe·HCl

H25920-06 H25920-14
AAH25920-06EA 46.9 USD
AAH25920-06 AAH25920-14
D-Phenylalanine methyl ester hydrochloride 99%
D-Phenylalanine methyl ester hydrochloride

Purity: 99% CAS No.: 13033-84-6 Molecular Formula: C10H13NO2.HCl Formula Weight: 215.68 Melting Point: 158-162° MDL No.: MFCD00066112

Formula: C₁₀H₁₃NO₂
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00066112
CAS Number: 13033-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR