3-Amino-4-phenyl-1H-pyrazole 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Phenyl-1H-pyrazol-3-amine

H25895-03 H25895-06
AAH25895-03EA 57.4 USD
AAH25895-03 AAH25895-06
3-Amino-4-phenyl-1H-pyrazole 95%
3-Amino-4-phenyl-1H-pyrazole

Purity: 95% CAS No.: 5591-70-8 Molecular Formula: C9H9N3 Formula Weight: 159.19 Melting Point: 170-173° MDL No.: MFCD01693715

Formula: C₉H₉N₃
MW: 159.19 g/mol
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD01693715
CAS Number: 5591-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR