Chlorobis(4-methylphenyl)phosphine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chlorodi(p-tolyl)phosphine
H25889-03
H25889-06
AAH25889-03EA
128
USD
AAH25889-03
AAH25889-06
Chlorobis(4-methylphenyl)phosphine 95%
Chlorobis(4-methylphenyl)phosphine
Purity: 95%
CAS No.: 1019-71-2
Molecular Formula: C14H14ClP
Formula Weight: 248.69
Boiling Point: 180-184°/10mm
UN No.: 1760
MDL No.: MFCD01630844
Formula:
C₁₄H₁₄ClP MW: 248.69 g/mol Boiling Pt: 180…184 °C/10 mm Density: 1.159 Storage Temperature: Ambient |
MDL Number:
MFCD01630844 CAS Number: 1019-71-2 UN: 1760 ADR: 8,II |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...