Chlorobis[4-(trifluoromethyl)phenyl]phosphine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H25888-03
H25888-06
AAH25888-03EA
124.52
USD
AAH25888-03
AAH25888-06
Chlorobis[4-(trifluoromethyl)phenyl]phosphine 97%
Chlorobis[4-(trifluoromethyl)phenyl]phosphine
Purity: 97%
CAS No.: 13685-24-0
Molecular Formula: C14H8ClF6P
Formula Weight: 356.64
Boiling Point: 110°/0.4mm
UN No.: 3265
MDL No.: MFCD04972304
Formula:
C₁₄H₈ClF₆P Boiling Pt: 110 °C/0.4 mm Storage Temperature: Ambient |
MDL Number:
MFCD04972304 CAS Number: 13685-24-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...