Chlorobis(4-methoxyphenyl)phosphine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H25887-03
H25887-06
AAH25887-03EA
126.69
USD
AAH25887-03
AAH25887-06
Chlorobis(4-methoxyphenyl)phosphine 98+%
Chlorobis(4-methoxyphenyl)phosphine
Purity: 98+%
CAS No.: 13685-30-8
Molecular Formula: C14H14ClO2P
Formula Weight: 280.69
Odor: Irritating
Melting Point: 61-64°
Boiling Point: 140°/1.5mm
UN No.: 3261
MDL No.: MFCD04972301
Formula:
C₁₄H₁₄ClO₂P MW: 280.69 g/mol Boiling Pt: 140 °C/1.5 mm Melting Pt: 61…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972301 CAS Number: 13685-30-8 UN: 3261 ADR: "8",III |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...