Chlorobis(3,5-dimethylphenyl)phosphine tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bis(3,5-dimethylphenyl)chlorophosphine

H25885-03 H25885-06
AAH25885-03EA 127 USD
AAH25885-03 AAH25885-06
Chlorobis(3,5-dimethylphenyl)phosphine tech. 90%, Technical Grade
Chlorobis(3,5-dimethylphenyl)phosphine

Purity: 90% CAS No.: 74289-57-9 Molecular Formula: C16H18ClP Formula Weight: 276.75 UN No.: 3261 MDL No.: MFCD01630841

Formula: C₁₆H₁₈ClP
MW: 276.75 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01630841
CAS Number: 74289-57-9
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR