Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine ≥98% may contain suspended dimethylamine hydrochloride crystals

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bis(3,5-bis(trifluoromethyl)phenyl)chlorophosphane

H25883-06 H25883-03
AAH25883-06EA 364.31 USD
AAH25883-06 AAH25883-03
Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine ≥98% may contain suspended dimethylamine hydrochloride crystals
Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine

Purity: 98+% CAS No.: 142421-57-6 Molecular Formula: C16H6ClF12P Formula Weight: 492.63 UN No.: 3265 MDL No.: MFCD01630852

Formula: C₁₆H₆ClF₁₂P
Storage Temperature: Ambient
MDL Number: MFCD01630852
CAS Number: 142421-57-6
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR