2-(Ethoxycarbonyl)phenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Ethoxycarbonyl)benzeneboronic acid, (2-(Ethoxycarbonyl)phenyl)boronic acid
H25867-06
H25867-03
AAH25867-06EA
83
USD
AAH25867-06
AAH25867-03
2-(Ethoxycarbonyl)phenylboronic acid 98%
2-(Ethoxycarbonyl)phenylboronic acid
Purity: 98%
CAS No.: 380430-53-5
Molecular Formula: C9H11BO4
Formula Weight: 193.99
Melting Point: 130-133°
MDL No.: MFCD02179453
Formula:
C₉H₁₁BO₄ MW: 193.99 g/mol Melting Pt: 130…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179453 CAS Number: 380430-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...