4-n-Propylpiperidine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Propylpiperidine

H25848-03 H25848-MD
AAH25848-03EA 162 USD
AAH25848-03 AAH25848-MD
4-n-Propylpiperidine 95%
4-n-Propylpiperidine

Purity: 95% CAS No.: 22398-09-0 Molecular Formula: C8H17N Formula Weight: 127.23 Flash Point: 48°(118°F) Refractive Index: 1.4530 Odor: Amine-like Boiling Point: 178-180° UN No.: 2733 MDL No.: MFCD00023151

Formula: C₈H₁₇N
MW: 127.23 g/mol
Boiling Pt: 178…180 °C
Density: 0.864
Flash Pt: 48 °C (118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00023151
CAS Number: 22398-09-0
EINECS: 244-956-9
UN: 2733
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR