Azetidin-3-ol hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Hydroxyazetidine hydrochloride
H25842-03
H25842-06
AAH25842-MDEA
35.4
USD
AAH25842-MD
AAH25842-03
AAH25842-06
Azetidin-3-ol hydrochloride 97%
Azetidin-3-ol hydrochloride
Purity: 97%
CAS No.: 18621-18-6
Molecular Formula: C3H7NO.HCl
Formula Weight: 109.56
Melting Point: 85-90°
MDL No.: MFCD02683887
Formula:
C₃H₈ClNO Melting Pt: 85…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683887 CAS Number: 18621-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...