N-Methyl-N-methoxydifluoroacetamide 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2-Difluoro-N-methoxy-N-methylacetamide
H25833-03
H25833-06
AAH25833-03EA
90.7
USD
AAH25833-03
AAH25833-06
AAH25833-MD
N-Methyl-N-methoxydifluoroacetamide 95%
N-Methyl-N-methoxydifluoroacetamide
Purity: 95%
CAS No.: 142492-01-1
Molecular Formula: C4H7F2NO2
Formula Weight: 139.10
Refractive Index: 1.4020
Boiling Point: 40-41°/0.7mm
MDL No.: MFCD06248152
Formula:
C₄H₇F₂NO₂ MW: 139.1 g/mol Boiling Pt: 40…41 °C/0.7 mm Storage Temperature: Ambient |
MDL Number:
MFCD06248152 CAS Number: 142492-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...