Cyclopropane-1,1-dicarboxylic acid mono methyl ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(Methoxycarbonyl)cyclopropanecarboxylic acid
H25828-03
H25828-06
AAH25828-03EA
28.7
USD
AAH25828-03
AAH25828-06
Cyclopropane-1,1-dicarboxylic acid mono methyl ester 97%
Cyclopropane-1,1-dicarboxylic acid mono methyl ester
Purity: 97%
CAS No.: 113020-21-6
Molecular Formula: C6H8O4
Formula Weight: 144.13
Flash Point:
Formula:
C₆H₈O₄ MW: 144.13 g/mol Boiling Pt: 85…87 °C/0.2 mm Melting Pt: 50…57 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD06800496 CAS Number: 113020-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...