3-(3,4-Dimethoxybenzoyl)propionic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(3,4-Dimethoxyphenyl)-4-oxobutanoic acid

H25806-14 H25806-06
AAH25806-14EA 331 USD
AAH25806-14 AAH25806-06
3-(3,4-Dimethoxybenzoyl)propionic acid 98%
3-(3,4-Dimethoxybenzoyl)propionic acid

Purity: 98% CAS No.: 5333-34-6 Molecular Formula: C12H14O5 Formula Weight: 238.24 Melting Point: 159-161° MDL No.: MFCD00033286

Formula: C₁₂H₁₄O₅
MW: 238.24 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00033286
CAS Number: 5333-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR