(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%

Supplier: Thermo Scientific Chemicals
Danger

H25784-MD H25784-03
AAH25784-MDEA 53.5 USD
AAH25784-MD AAH25784-03
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene

Purity: 97% CAS No.: 18531-95-8 Molecular Formula: C20H16N2 Formula Weight: 284.36 Melting Point: 236-242° Beilstein Reference: 2419071 Optical Rotation: -150° (c=1 in pyridine) MDL No.: MFCD00145204

Formula: C₂₀H₁₆N₂
Melting Pt: 236…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00145204
CAS Number: 18531-95-8
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR