3,3-Dimethylpiperidine 95%

Supplier: Thermo Scientific Chemicals
Danger

H25753-03 H25753-MD
AAH25753-03EA 178 USD
AAH25753-03 AAH25753-MD
3,3-Dimethylpiperidine 95%
3,3-Dimethylpiperidine

Purity: 95% CAS No.: 1193-12-0 Molecular Formula: C7H15N Formula Weight: 113.20 Flash Point: 26°(79°F) Refractive Index: 1.4475 Odor: Amine-like Boiling Point: 134-136° UN No.: 2733 MDL No.: MFCD00005995

Formula: C₇H₁₅N
MW: 113.20 g/mol
Boiling Pt: 134…136 °C
Density: 0.81
Flash Pt: 26 °C(79 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005995
CAS Number: 1193-12-0
EINECS: 214-767-6
UN: 2733
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR