Ethyl-2,4-dioxo-5-methylhexanoate 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 5-methyl-2,4-dioxohexanoate
B25735-06
B25735-14
AAB25735-06EA
52.3
USD
AAB25735-06
AAB25735-14
AAB25735-03
Ethyl-2,4-dioxo-5-methylhexanoate 95%
Ethyl-2,4-dioxo-5-methylhexanoate
Purity: 95%
CAS No.: 64195-85-3
Molecular Formula: C9H14O4
Formula Weight: 186.21
MDL No.: MFCD07323638
Formula:
C₉H₁₄O₄ MW: 186.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07323638 CAS Number: 64195-85-3 EINECS: 264-728-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...