6-Chloro-7-deazapurine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-7H-pyrrolo[2,3-d]pyrimidine

B25713-03 B25713-06
AAB25713-03EA 208 USD
AAB25713-03 AAB25713-06
6-Chloro-7-deazapurine 98%
6-Chloro-7-deazapurine

Purity: 98% CAS No.: 3680-69-1 Molecular Formula: C6H4ClN3 Formula Weight: 153.52 Storage Temperature: 2°C to 8°C Melting Point: 190-192° MDL No.: MFCD01686865

Formula: C₆H₄ClN₃
MW: 153.57 g/mol
Melting Pt: 190…192 °C
Storage Temperature: Ambient
MDL Number: MFCD01686865
CAS Number: 3680-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR