7-Bromo-1,2,3,4-tetrahydroisoquinoline 97%

Supplier: Thermo Scientific Chemicals
Warning

B25712-06 B25712-03
AAB25712-06EA 290 USD
AAB25712-06 AAB25712-03
7-Bromo-1,2,3,4-tetrahydroisoquinoline 97%
7-Bromo-1,2,3,4-tetrahydroisoquinoline

Purity: 97% CAS No.: 17680-55-6 Molecular Formula: C9H10BrN Formula Weight: 212.09 Melting Point: 32-35° MDL No.: MFCD06739047

Formula: C₉H₁₀BrN
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD06739047
CAS Number: 17680-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR