Methyl-4-fluoro-3-hydroxybenzoate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 4-fluoro-3-hydroxybenzoate
B25683-14
B25683-06
AAB25683-14EA
245
USD
AAB25683-14
AAB25683-06
Methyl-4-fluoro-3-hydroxybenzoate 98+%
Methyl 4-fluoro-3-hydroxybenzoate
Purity: 98+%
CAS No.: 214822-96-5
Molecular Formula: C8H7FO3
Formula Weight: 170.14
Melting Point: 89-91°
MDL No.: MFCD06797457
Formula:
C₈H₇FO₃ MW: 170.14 g/mol Melting Pt: 89…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797457 CAS Number: 214822-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...