4-Bromo-1-chloro-2-ethylbenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25613-14
B25613-06
AAB25613-14EA
200
USD
AAB25613-14
AAB25613-06
4-Bromo-1-chloro-2-ethylbenzene 98+%
4-Bromo-1-chloro-2-ethylbenzene
Purity: 98+%
CAS No.: 289039-22-1
Molecular Formula: C8H8BrCl
Formula Weight: 219.51
MDL No.: MFCD00672954
Formula:
C₈H₈BrCl MW: 219.51 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00672954 CAS Number: 289039-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...