Ethyl-4-hydroxycyclohexanecarboxylate cis- and trans-mixture 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 4-hydroxycyclohexanecarboxylate
B25572-06
B25572-14
AAB25572-06EA
63.9
USD
AAB25572-06
AAB25572-14
Ethyl-4-hydroxycyclohexanecarboxylate cis- and trans-mixture 97%
Ethyl-4-hydroxycyclohexanecarboxylate
Purity: 97%
CAS No.: 17159-80-7
Molecular Formula: C9H16O3
Formula Weight: 172.22
Flash Point:
Formula:
C₉H₁₆O₃ MW: 172.22 g/mol Boiling Pt: 127…134 °C/0.1 mm Density: 1.068 Flash Pt: >110(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00192156 CAS Number: 17159-80-7 EINECS: 241-215-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...