Amberlite® IRA-402 (Cl), Amberlite™ ion exchange resin

Supplier: Thermo Scientific Chemicals

B25556-22 B25556-36 B25556-0E
AAB25556-22EA 24.35 USD
AAB25556-22 AAB25556-36 AAB25556-0E
Amberlite® IRA-402 (Cl), Amberlite™ ion exchange resin
Amberlite® IRA-402 (Cl)

CAS No.: 52439-77-7 MDL No.: MFCD01864002

Density: 1.07
Storage Temperature: Ambient
MDL Number: MFCD00145564
CAS Number: 52439-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR