Methyl-4-iodo-2-methoxybenzoate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25529-06
B25529-14
AAB25529-06EA
74
USD
AAB25529-06
AAB25529-14
Methyl-4-iodo-2-methoxybenzoate 98+%
Methyl-4-iodo-2-methoxybenzoate
Purity: 98+%
CAS No.: 148490-97-5
Molecular Formula: C9H9IO3
Formula Weight: 292.07
Melting Point: 44-46°
MDL No.: MFCD06797865
Formula:
C₉H₉IO₃ Melting Pt: 44…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052925 CAS Number: 35387-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...