4-Bromo-2,5-dimethoxybenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

B25500-06 B25500-14
AAB25500-06EA 88.7 USD
AAB25500-06 AAB25500-14
4-Bromo-2,5-dimethoxybenzaldehyde 98+%
4-Bromo-2,5-dimethoxybenzaldehyde

Purity: 98+% CAS No.: 31558-41-5 Molecular Formula: C9H9BrO3 Formula Weight: 245.08 Melting Point: 132-135° MDL No.: MFCD00156964

Formula: C₉H₉BrO₃
MW: 245.08 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00156964
CAS Number: 31558-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR