2-Bromo-1,3-dimethoxybenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25485-03
B25485-06
B25485-14
AAB25485-03EA
23.3
USD
AAB25485-03
AAB25485-06
AAB25485-14
2-Bromo-1,3-dimethoxybenzene 98%
2-Bromo-1,3-dimethoxybenzene
Purity: 98%
CAS No.: 16932-45-9
Molecular Formula: C8H9BrO2
Formula Weight: 217.06
Melting Point: 91-94°
MDL No.: MFCD07780177
Formula:
C₈H₉BrO₂ MW: 217.06 g/mol Melting Pt: 91…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD07780177 CAS Number: 16932-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...