3-Aminopropionitrile fumarate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25440-06
B25440-14
AAAB25440-06EA
54.9
USD
AAAB25440-06
AAAB25440-14
3-Aminopropionitrile fumarate 98+%
3-Aminopropionitrile fumarate
Purity: 98+%
CAS No.: 2079-89-2
Molecular Formula: (NH2CH2CH2CN)2.HO2CCH=CHCO2H
Formula Weight: 256.26
Odor: Odorless
Melting Point: ca 177° dec.
Beilstein Reference: 3573532
MDL No.: MFCD00078321
Formula:
C₁₀H₁₆N₄O₄ MW: 256.26 g/mol Melting Pt: ca 177 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013142 CAS Number: 2079-89-2 EINECS: 218-208-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...