Methyl-4-iodsalicylate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 2-hydroxy-4-iodobenzoate
B25438-06
B25438-14
AAB25438-06EA
47.6
USD
AAB25438-06
AAB25438-14
Methyl-4-iodsalicylate 98%
Methyl 4-iodsalicylate
Purity: 98%
CAS No.: 18179-39-0
Molecular Formula: C8H7IO3
Formula Weight: 278.05
Melting Point: 68-72°
MDL No.: MFCD06797864
Formula:
C₈H₇IO₃ Melting Pt: 68…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797864 CAS Number: 18179-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...