Papaverine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

B25412-14 B25412-06
AAAB25412-14EA 131 USD
AAAB25412-14 AAAB25412-06
Papaverine hydrochloride 99%
Papaverine hydrochloride

A phosphodiesterase inhibitor


Purity: 99% CAS No.: 61-25-6 Molecular Formula: C20H21NO4.HCl Formula Weight: 375.85 Melting Point: ca 220° dec. MDL No.: MFCD00012745

Formula: C₂₀H₂₁NO₄·HCl
Melting Pt: ca 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012745
CAS Number: 61-25-6
EINECS: 200-502-1
UN: 2811
ADR: "6.1",III
Merck Index: 14,7019

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR