Cyclobutylamine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cyclobutanamine hydrochloride
B25400-14
B25400-06
AAB25400-14EA
330
USD
AAB25400-14
AAB25400-06
Cyclobutylamine hydrochloride 97%
Cyclobutylamine hydrochloride
Purity: 97%
CAS No.: 6291-01-6
Molecular Formula: C4H9N.HCl
Formula Weight: 107.58
Melting Point: 180-183°
MDL No.: MFCD00034953
Formula:
C₄H₉N·HCl MW: 107.58 g/mol Melting Pt: 180…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00034953 CAS Number: 6291-01-6 EINECS: 228-540-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...