4-Phenoxybenzonitrile 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25364-04
B25364-09
AAAB25364-04EA
28.3
USD
AAAB25364-04
AAAB25364-09
4-Phenoxybenzonitrile 96%
4-Phenoxybenzonitrile
Purity: 96%
CAS No.: 3096-81-9
Molecular Formula: C13H9NO
Formula Weight: 195.22
Flash Point:
Formula:
C₁₃H₉NO MW: 195.22 g/mol Melting Pt: 46…48 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017346 CAS Number: 3096-81-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...