Chlorpheniramine maleate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Chlorphenamine maleate
B25238-14
B25238-06
AAB25238-14EA
79.9
USD
AAB25238-14
AAB25238-06
Chlorpheniramine maleate 99%
Chlorpheniramine maleate
An antihistaminic agent
Purity: 99%
CAS No.: 113-92-8
Molecular Formula: C20H23ClN2O4
Formula Weight: 390.86
Melting Point: 130-135°
MDL No.: MFCD00069225
Formula:
C₁₆H₁₉ClN₂ MW: 390.86 g/mol Melting Pt: 130…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00069225 CAS Number: 113-92-8 EINECS: 204-037-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...