1,3-Di-o-tolylguanidine 99%

Supplier: Thermo Scientific Chemicals
Danger

B25127-36 B25127-22
AAAB25127-36EA 237 USD
AAAB25127-36 AAAB25127-22
1,3-Di-o-tolylguanidine 99%
1,3-Di-o-tolylguanidine

Purity: 99% CAS No.: 97-39-2 Molecular Formula: C15H17N3 Formula Weight: 239.32 Melting Point: 176-178° UN No.: 2811 MDL No.: MFCD00008513

Formula: C₁₅H₁₇N₃
MW: 239.32 g/mol
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00008513
CAS Number: 97-39-2
EINECS: 202-577-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR