Methyl-6-bromo-2-naphthoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 6-bromo-2-naphthoate

B25106-14 B25106-06
AAAB25106-03EA 22.75 USD
AAAB25106-03 AAAB25106-14 AAAB25106-06
Methyl-6-bromo-2-naphthoate 98%
Methyl 6-bromo-2-naphthoate

Purity: 98% CAS No.: 33626-98-1 Molecular Formula: C12H9BrO2 Formula Weight: 265.11 Melting Point: 121-123° Beilstein Reference: 2578943

Formula: C₁₂H₉BrO₂
MW: 265.11 g/mol
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00100408
CAS Number: 33626-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR