4-Benzyloxy-2-hydroxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Benzyloxy-2(1H)-pyridone

B24774-06 B24774-14
AAAB24774-06EA 112 USD
AAAB24774-06 AAAB24774-14 AAAB24774-03
4-Benzyloxy-2-hydroxypyridine 97%
4-Benzyloxy-2-hydroxypyridine

Purity: 97% CAS No.: 53937-02-3 Molecular Formula: C12H11NO2 Formula Weight: 201.22 Melting Point: 194-198° MDL No.: MFCD04039771

Formula: C₁₂H₁₁NO₂
MW: 201.22 g/mol
Melting Pt: 194…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00191783
CAS Number: 53937-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR