4-Methylphenylacetone 97+%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(p-Tolyl)propan-2-one

B24728-03 B24728-06
AAAB24728-03EA 47.6 USD
AAAB24728-03 AAAB24728-06
4-Methylphenylacetone 97+%
4-Methylphenylacetone

Purity: 97+% CAS No.: 2096-86-8 Molecular Formula: C10H12O Formula Weight: 148.20 MDL No.: MFCD00017250

Formula: C₁₀H₁₂O
MW: 148.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017250
CAS Number: 2096-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR