Methyl diphenylacetate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl Diphenylacetate

B24694-22 B24694-06 B24694-14
AAAB24694-22EA 529 USD
AAAB24694-22 AAAB24694-06 AAAB24694-14
Methyl diphenylacetate 98%
Methyl diphenylacetate

Purity: 98% CAS No.: 3469-00-9 Molecular Formula: (C6H5)2CHCO2CH3 Formula Weight: 226.28 Melting Point: 59-61° MDL No.: MFCD00025869

Formula: C₁₅H₁₄O₂
MW: 226.28 g/mol
Melting Pt: 59…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00025869
CAS Number: 3469-00-9
EINECS: 222-431-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR