Methyl-4-hydroxy-2-hexynoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24690-03
B24690-06
AAAB24690-03EA
104
USD
AAAB24690-03
AAAB24690-06
AAAB24690-14
Methyl-4-hydroxy-2-hexynoate 97%
Methyl-4-hydroxy-2-hexynoate
Purity: 97%
CAS No.: 112780-04-8
Molecular Formula: C7H10O3
Formula Weight: 142.15
Refractive Index: 1.4620
MDL No.: MFCD04039770
Formula:
C₇H₁₀O₃ MW: 142.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04039770 CAS Number: 112780-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...