Potassium sulfamate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Aminosulphonic acid Potassium Salt, sulphamic acid Potassium Salt
B24447-14
B24447-22
B24447-36
AAB24447-14EA
76.35
USD
AAB24447-14
AAB24447-22
AAB24447-36
Potassium sulfamate 98%
Potassium sulfamate
Purity: 98%
CAS No.: 13823-50-2
Molecular Formula: H2KNO3S
Formula Weight: 135.18
Physical Form: Cryst solid
Sensitivity: Hygroscopic
Appearance: White
MDL No.: MFCD00059941
Formula:
H₃NO₃S MW: 135.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059941 CAS Number: 13823-50-2 EINECS: 237-515-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...