Benzhydrylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α-Phenylbenzylamine, α-Aminodiphenylmethane, Diphenylmethanamine, Aminodiphenylmethane, 1,1-Diphenylmethanamine, Benzhydrylamin

B24303-14 B24303-22
AAB24303-14EA 72 USD
AAB24303-14 AAB24303-22
Benzhydrylamine 97%
Benzhydrylamine

Slightly soluble in water


Purity: 97% CAS No.: 91-00-9 Molecular Formula: C13H13N Formula Weight: 183.25 Flash Point: 110°(230°F) Refractive Index: 1.5950 Melting Point: 11-13° Boiling Point: 294-296° Solubility: Slightly soluble in water Beilstein Reference: 776434 Merck Reference: 14,1076 UN No.: 2735 MDL No.: MFCD00008059

Formula: C₁₃H₁₃N
Boiling Pt: 294…296 °C
Melting Pt: 11…13 °C
Density: 1.064
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008059
CAS Number: 91-00-9
EINECS: 202-032-2
ADR: 8,III
Merck Index: 14,1076

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR