Cobalt(II) gluconate hydrate

Supplier: Thermo Scientific Chemicals
Warning

B24293-14 B24293-22
AAB24293-14EA 50.3 USD
AAB24293-14 AAB24293-22
Cobalt(II) gluconate hydrate
Cobalt(II) gluconate hydrate

CAS No.: 71957-08-9 Molecular Formula: C12H22CoO14.xH2O Formula Weight: 449.23(anhy) Melting Point: 300° MDL No.: MFCD02684006

Formula: C₁₂H₂₂CoO₁₄·xH₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD02684006
CAS Number: 1809162-99-9
EINECS: 276-206-1
UN: 3288
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR