Triethylbutylammonium bromide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(1-Butyl)triethylammonium bromide
B24128-22
B24128-14
AAB24128-22EA
155
USD
AAB24128-22
AAAB24128-14
Triethylbutylammonium bromide 97%
Triethylbutylammonium bromide
Purity: 97%
CAS No.: 13028-69-8
Molecular Formula: CH3(CH2)3N(CH2CH3)3Br
Formula Weight: 238.21
Melting Point: 212-215°
MDL No.: MFCD00055541
Formula:
C₁₀H₂₄BrN Melting Pt: 212…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055541 CAS Number: 13028-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...