Methyl-4-formylbenzoate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl terephthalaldehydate, Methyl 4-formylbenzoate
B24091-06
B24091-14
B24091-22
AAB24091-06EA
50.2
USD
AAB24091-06
AAB24091-14
AAB24091-22
Methyl-4-formylbenzoate 98+%
Methyl-4-formylbenzoate
Purity: 98+%
CAS No.: 1571-08-0
Molecular Formula: C9H8O3
Formula Weight: 164.16
Flash Point: 128°(262°F)
Melting Point: 59-63°
Boiling Point: 264-266°
Beilstein Reference: 775895
MDL No.: MFCD00006950
Formula:
C₉H₈O₃ MW: 164.16 g/mol Boiling Pt: 264…266 °C Melting Pt: 59…63 °C Density: 1.200 Flash Pt: 128 °C(262 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006950 CAS Number: 1571-08-0 EINECS: 216-385-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...