5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
5(4)-Amino-4(5)-imidazolecarboxamide Hydrochloride, AICA, 5(4)-Amino-4(5)-(aminocarbonyl)imidazole Hydrochloride, 4-Aminoimidazole-5-carboxamide hydrochloride, 4-Amino-5-imidazolecarboxamide hydrochloride, 5-Amino-4-imidazolecarboxamide hydrochloride
B24012-06
B24012-14
AAB24012-06EA
82.9
USD
AAB24012-06
AAB24012-03
AAB24012-14
5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%
5-Amino-1H-imidazole-4-carboxamide hydrochloride
Purity: 98%
CAS No.: 72-40-2
Molecular Formula: C4H6N4O.HCl
Formula Weight: 162.58
Odor: Odorless
Melting Point: ca 270° dec.
Beilstein Reference: 3701645
MDL No.: MFCD00012704
Formula:
C₄H₇ClN₄O MW: 162.58 g/mol Melting Pt: ∼ 270 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012704 CAS Number: 72-40-2 EINECS: 200-778-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...