N-Acetyl-L-phenylalanine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Ac-Phe-OH

B23812-14 B23812-03 B23812-06
AAB23812-14EA 293 USD
AAB23812-14 AAB23812-03 AAB23812-06
N-Acetyl-L-phenylalanine 99%
N-Acetyl-L-phenylalanine

Purity: 99% CAS No.: 2018-61-3 Molecular Formula: C11H13NO3 Formula Weight: 207.23 Melting Point: 166-170° Optical Rotation: +40° (c=1 in methanol) MDL No.: MFCD00063158

Formula: C₁₁H₁₃NO₃
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00063158
CAS Number: 2018-61-3
EINECS: 217-959-8

Order Now

Specification Test Results

Optical rotation +40° (c=1 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR