Ammonium 1-pyrrolidinecarbodithioate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD

B23731-14 B23731-22
AAB23731-14EA 127.59 USD
AAB23731-14 AAB23731-22
Ammonium 1-pyrrolidinecarbodithioate 98%
Ammonium 1-pyrrolidinecarbodithioate

Purity: 98% CAS No.: 5108-96-3 Molecular Formula: C5H12N2S2 Formula Weight: 164.29 Melting Point: 150-155° Beilstein Reference: 3730472

Formula: C₅H₁₂N₂S₂
MW: 146.3 · 18.0 g/mol
Melting Pt: 150…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00012720
CAS Number: 5108-96-3
EINECS: 225-834-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR